Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Thermo Scientific™ BioBasic™ 4 KAPPA 5µm Capillary HPLC Columns

Thermo Scientific™ BioBasic™ 4 KAPPA 5µm Capillary HPLC Columns operate at the low flowrates required for greater ionization efficiencies.

Specifications

Carbon Load 4%
For Use With LC/MS, APCI, ESI
Stationary Phase C4
USP Type L26
Column Type Reversed Phase
View More Specs

Products 6
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
72305050265
View Documents
Thermo Scientific™
72305050265
5 μm 0.18 mm 50 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
72305050365
View Documents
Thermo Scientific™
72305050365
5 μm 0.32 mm 50 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
72305050565
View Documents
Thermo Scientific™
72305050565
5 μm 0.5 mm 50 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
72305100565
View Documents
Thermo Scientific™
72305100565
5 μm 0.5 mm 100 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
72305150365
View Documents
Thermo Scientific™
72305150365
5 μm 0.32 mm 150 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
72305150565
View Documents
Thermo Scientific™
72305150565
5 μm 0.5 mm 150 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Thermo Scientific™ BioBasic™ 4 KAPPA 5µm Capillary HPLC Columns operate at the low flowrates required for greater ionization efficiencies. Highly efficient KAPPA capillary columns meet the sensitivity needs of demanding LC/MS applications, particularly proteomics.

  • Rugged sleeved design for robustness
  • Excellent for LC/MS analysis
  • Particle size: 5µm

Features of BioBasic 4 Solid Phase

  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides an proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Specifications

Specifications

4%
C4
Reversed Phase
5 μm
2 to 10
LC/MS, APCI, ESI
L26
Yes
300 Å
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit