Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ 4 KAPPA Capillary Guard Columns

Protect Thermo Scientific™ BioBasic™ 4 KAPPA Capillary HPLC Columns from sample debris and interfering substances.

Specifications

Carbon Load 4%
Endcapped Yes
Pore Size 300Å
USP Type L26
Length (Metric) 30 mm
View More Specs

Products 2
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
72305030215
View Documents
Thermo Scientific™
72305030215
5 µm 0.18 mm 30 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
72305030315
View Documents
Thermo Scientific™
72305030315
5 µm 0.32 mm 30 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Protect your analytical HPLC column from damage caused by sample debris and interfering substances. It's the economical way to extend the lifetime of your analytical column with minimal impact on performance.

Use Thermo Scientific™ BioBasic™ 4 KAPPA Capillary Guard Columns with BioBasic 4 KAPPA Capillary HPLC Columns, designed to operate at low flow rates required for greater ionization efficiencies and meet the sensitivity needs of demanding LC/MS applications, particularly proteomics.

Features of BioBasic 4 Solid Phase

  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides an proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Specifications

Specifications

4%
300Å
30 mm
2 to 10
Yes
L26
5 µm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit