Learn More
Thermo Scientific™ BioBasic™ 4 LC Columns
BioBasic 4 LC Columns are designed for the analysis of larger proteins and peptides.
Manufacturer:  Thermo Scientific™ 72305151030
 View more versions of this product
Catalog No. 03-050-872
Add to cart
Description
Description
Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.
  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.
Specifications
Specifications
4%
Reversed Phase
Yes
400bar
5 µm
300 Å
60°C
L26
Analytical Column
1 mm
150 mm
Silica, Spherical Fully Porous Ultrapure
2 to 9
100 m²/g
Analytical Column
C4
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.