Learn More
Thermo Scientific™ BioBasic™ 4 LC Columns Available on GSA/VA Contract for Federal Government customers only.
BioBasic 4 LC Columns are designed for the analysis of larger proteins and peptides.
Manufacturer:  Thermo Scientific™ 72305052130
 View more versions of this product
Catalog No. 72305052130
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.
  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.
Description & Specifications
Specifications
4%
Reversed Phase
Yes
400bar
5μm
300Å
60°C
L26
Analytical Column
2.1mm
50mm
Silica, Spherical Fully Porous Ultrapure
2 to 9
100m²/g
Analytical Column
C4