Learn More
Thermo Scientific™ BioBasic™ 8 HPLC Columns
Use this column for reversed-phase separation of proteins and other biomolecules.
Supplier:  Thermo Scientific™ 72205014001
 View more versions of this product
Catalog No. 03-051-671
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.
  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Guard Cartridge, 4 Pk.
Uniguard Direct-Connection and Stand Alone Holder
5800 psi (400 bar)
2 to 9
100 m²/g
L7
BioBasic
5%
Yes
Guard Cartridge
4 mm
10 mm
5 μm
300 Å
60°C
C8
Spherical, Fully Porous, Ultrapure Silica
Reversed Phase
Reversed Phase
Product Suggestions
Product Suggestions