Learn More
Thermo Scientific™ BioBasic™ 8 LC Columns Available on GSA/VA Contract for Federal Government customers only.
Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.
Manufacturer:  Thermo Scientific™ 72205011001
 View more versions of this product
Catalog No. 03-051-667
  Check Availability
Add to cart
Specifications
Description
Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.
  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call Customer Service for more information.
Description & Specifications
Specifications
5%
Reversed Phase
Yes
10mm
5μm
300Å
60°C
L7
Guard Cartridge, 4 PK
1mm
Uniguard Direct-connection and Stand Alone Holder
400bar
2 to 9
100m²/g
Guard Cartridge, 4 PK