Learn More
Thermo Scientific™ BioBasic™ 8 LC Columns Available on GSA/VA Contract for Federal Government customers only.
Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.
Manufacturer:  Thermo Scientific™ 72205012101
 View more versions of this product
Catalog No. 03-051-668
Add to cart
Description
Description
Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.
  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call Customer Service for more information.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
5%
Reversed Phase
Yes
10 mm
5 µm
300 Å
60°C
L7
Guard Cartridge, 4 Pk.
2.1 mm
Uniguard Direct-connection and Stand Alone Holder
400bar
2 to 9
100 m²/g
Guard Cartridge, 4 Pk.