Thermo Scientific™ BioBasic™ 8 LC Columns Available on GSA/VA Contract for Federal Government customers only.

Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.

Manufacturer: Thermo Scientific™ 72205152130

 View more versions of this product

Catalog No. 03-051-692

  Check Availability
Add to Cart

    Specifications

    Column Format Analytical Column
    Diameter (Metric) 2.1mm
    Stationary Phase BioBasic 8
    USP Type L7
    Column Type Reversed Phase
    Max. Pressure 400bar
    Packing Material Spherical, fully porous ultrapure silica
    Particle Size 5µm
    Temperature 60°C
    Pore Size 300Å
    pH 2 - 10
    For Use With LC/MS
    Surface Area 100m²/g
    Type Analytical Column
    Length (Metric) 150mm
    Carbon Load 5%
    Endcapped Yes

    Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.

    • 300Å pore size for improved biomolecule analysis
    • An excellent starting column for protein and peptide analysis
    • Outstanding reproducibility, efficiency and column lifetime
    • Excellent for LC/MS separations

    Ordering Information:

    Other column dimensions are available, including preparative columns. Please call Customer Service for more information.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit