Learn More
BioBasic™ 8 LC Columns
Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.
Manufacturer:  Thermo Scientific 72205104630
 View more versions of this product
Catalog No. 03-050-866
  Check Availability
Add to cart
Description
Description
Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.
  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call Customer Service for more information.
Specifications
Specifications
5%
Reversed Phase
Yes
400bar
5μm
300Å
60°C
L7
Analytical Column
4.6mm
100mm
Silica, Spherical Fully Porous Ultrapure
2 to 9
100m²/g
Analytical Column
C8