Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ 8 PEEK Bio-Inert HPLC Columns, 5µm Particle Size

Thermo Scientific™ BioBasic 8 PEEK Bio-Inert HPLC Columns, 5µm Particle Size, are optimized for the separation of a wide range of peptides.

Manufacturer:  Thermo Scientific™ 72205252168

 View more versions of this product

Catalog No. 03-051-696


Add to cart

Description

Description

Thermo Scientific™ BioBasic 8 PEEK Bio-Inert HPLC Columns, 5µm Particle Size, are optimized for the separation of a wide range of peptides. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules. Optimized for the separation of medium hydrophobicity peptides, nominally less than 20,000 daltons.

  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available in bio-inert column hardware. Please call Customer Service for more information.

Specifications

Specifications

Reversed Phase
5%
Yes
2.1 mm
250 mm
2 to 9
100 m²/g
C8
Silica
Reversed Phase
PEEK Column
LC/MS
5 µm
300 Å
L7
BioBasic
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit