Thermo Scientific™ BioBasic™ AX LC Columns Available on GSA/VA Contract for Federal Government customers only.

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

Manufacturer: Thermo Scientific™ 73105-014001

 View more versions of this product

Catalog No. 73105014001

  Check Availability
Add to Cart

    Specifications

    Column Type Ion Exchange
    Type Guard Cartridge, 4 PK
    Carbon Load 3%
    USP Type N/A
    Endcapped No
    Max. Pressure 400bar
    Particle Size 5µm
    Temperature 60°C
    Pore Size 300Å
    pH 2 to 8
    Stationary Phase Polyethyleneimine
    Surface Area 100m²/g
    Length (Metric) 10mm
    Technique Anion Exchange
    Column Format Guard Cartridge, 4 PK
    Diameter (Metric) 4mm

    Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

    • 5µm particle size, 300Å pore size
    • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
    • Low hydrophobicity
    • Capacity of 0.22mEq/g; 3% carbon load
    • Use for normal phase and hydrophilic interaction chromatography (HILIC)
    • Stable under demanding pH conditions (2 to 8 pH range)

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit