Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ AX LC Columns Available on GSA/VA Contract for Federal Government customers only.

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

Manufacturer:  Thermo Scientific™ 73105102130

 View more versions of this product

Catalog No. 73105102130


  Check Availability
Add to cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


Specifications

Description

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

  • 5µm particle size, 300Å pore size
  • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
  • Low hydrophobicity
  • Capacity of 0.22mEq/g; 3% carbon load
  • Use for normal phase and hydrophilic interaction chromatography (HILIC)
  • Stable under demanding pH conditions (2 to 8 pH range)
Description & Specifications

Specifications

3%
Ion Exchange
No
100mm
Silica, Spherical Fully Porous Ultrapure
2 to 8
100m²/g
N/A
Polyethyleneimine
Analytical Column
2.1mm
Low
400bar
5μm
300Å
60°C
Anion Exchange
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit