Thermo Scientific™ BioBasic™ AX LC Columns Available on GSA/VA Contract for Federal Government customers only.

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

Manufacturer: Thermo Scientific™ 73105053030

 View more versions of this product

Catalog No. 73105053030

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Column Format Analytical Column
    Diameter (Metric) 3mm
    For Use With Normal Phase; HILIC Chromatography
    Particle Size 5µm
    Pore Size 300Å
    pH 2-8
    Carbon Load 3%
    Surface Area 100m²/g
    Length (Metric) 50mm
    Anion Exchange Capacity 0.22mEq/g
    Stationary Phase BioBasic AX
    Hydrophobicity Low
    USP Type N/A
    Endcapped No
    Column Type Ion Exchange
    Max. Pressure 400bar
    Packing Material Spherical, fully porous ultrapure silica
    Temperature 60°C
    Technique Anion Exchange

    Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

    • 5µm particle size, 300Å pore size
    • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
    • Low hydrophobicity
    • Capacity of 0.22mEq/g; 3% carbon load
    • Use for normal phase and hydrophilic interaction chromatography (HILIC)
    • Stable under demanding pH conditions (2 to 8 pH range)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit