Thermo Scientific™ BioBasic™ SCX LC Columns Available on GSA/VA Contract for Federal Government customers only.

BioBasic SCX LC Columns are optimized for the separation of proteins, peptides and other cationic species.

Manufacturer: Thermo Scientific™ 73205252130

 View more versions of this product

Catalog No. 73205252130

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Column Format Analytical Column
    Diameter (Metric) 2.1mm
    Column Type Cation Exchange
    Max. Pressure 400bar
    Packing Material Spherical, fully porous ultrapure silica
    Particle Size 5µm
    Pore Size 300Å
    pH 2 to 8
    For Use With LC/MS, APCI, ESI
    Stationary Phase BioBasic SCX
    Type Analytical Column
    Length (Metric) 250mm
    Carbon Load 3%
    USP Type L52
    Endcapped No
    Temperature 60°C
    Surface Area 100m²/g

    Use Thermo Scientific™ BioBasic™ SCX LC Columns for the optimized separation of proteins, peptides and other cationic species. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules with higher efficiency than polymeric ion exchange.

    • Strong cation exchange phase based on sulfonic acid chemistry
    • Separation and retention of basic and other cationic species
    • 300Å pore size for enhanced protein and peptide separations
    • Outstanding stability under demanding pH conditions

    Ordering Information:

    Other column dimensions are available. Please call Customer Service for more information.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit