Learn More
Thermo Scientific™ BioBasic™ SCX LC Columns
BioBasic SCX LC Columns are optimized for the separation of proteins, peptides and other cationic species.
Manufacturer:  Thermo Scientific™ 73205103030
 View more versions of this product
Catalog No. 03-050-886
Add to cart
Description
Description
Use Thermo Scientific™ BioBasic™ SCX LC Columns for the optimized separation of proteins, peptides and other cationic species. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules with higher efficiency than polymeric ion exchange.
  • Strong cation exchange phase based on sulfonic acid chemistry
  • Separation and retention of basic and other cationic species
  • 300Å pore size for enhanced protein and peptide separations
  • Outstanding stability under demanding pH conditions
Ordering Information:
Other column dimensions are available. Please call Customer Service for more information.
Specifications
Specifications
3%
Ion Exchange
No
400bar
5 µm
300 Å
60°C
L52
Analytical Column
3 mm
100 mm
Silica, Spherical Fully Porous Ultrapure
2 to 8
100 m²/g
Analytical Column
Sulfonic acid
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.