Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ SEC 60 LC Columns Available on GSA/VA Contract for Federal Government customers only.

Outstanding size separation of water soluble compounds.

Manufacturer:  Thermo Scientific™ 73305307846

 View more versions of this product

Catalog No. 03-170-385


  Check Availability
Add to cart

Specifications

Description

Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 60 LC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 60Å pore size
  • Particle size: 5µm

Ordering Information:

For information on bulk quantities, please inquire.

Description & Specifications

Specifications

High Temperature Column
7.8mm
300mm
Silica, Spherical Fully Porous Ultrapure
60Å
L33
Size Exclusion
Size Exclusion Chromatography
400bar
5μm
High Temperature Column
Diol
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit