Learn More
BioBasic™ SEC 60 LC Columns
Outstanding size separation of water soluble compounds.
Manufacturer:  Thermo Scientific 73305037821
 View more versions of this product
Catalog No. 03-050-891
  Check Availability
Add to cart
Description
Description
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 60 LC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.
  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 60Å pore size
  • Particle size: 5µm
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
Specifications
Guard Column
7.8mm
30mm
5μm
Guard Column
Diol
Size Exclusion
Size Exclusion Chromatography
400bar
60Å
L33