Learn More
Thermo Scientific™ BioBasic™ SEC 120 LC Columns
Outstanding size separation of water soluble compounds.
Manufacturer:  Thermo Scientific™ 73405037821
 View more versions of this product
Catalog No. 03-050-893
Add to cart
Description
Description
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 120 LC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.
  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 120Å pore size
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
Specifications
Guard Column
7.8 mm
30 mm
5 µm
Guard Column
Diol
Size Exclusion
Size Exclusion Chromatography
400bar
120 Å
L33
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.