Learn More
20 20 Genesystems IncSupplier Diversity Partner BIOCHECK KIT 20/20 25/CS  DFS Item
Manufacturer:  20 20 Genesystems IncSupplier Diversity Partner GB1001CS
The BioCheck Powder Screening Test Kit is used by first responders as a first step to investigate suspicious powders for bioterrorism agents. The Kit works by quickly identifying the presence or absence of a biomolecule found in all living materials and many toxins. It, therefore, provides a rapid screen for the possible presence of bioterrorism agents such as anthrax and ricin toxin, while ruling out most of the ordinary substances that citizens have frequently feared to be possible weapons of mass destruction (WMD). Case of 25.
Catalog No. NC0441806