Learn More
BioCision™ Automation-Friendly Sample Modules for CoolBox™ XT/2XT Systems
Thermo-conductive tube holders with SBS-footprint modules for easy transfer to robotic platforms
Manufacturer:  BioCision™ BCS534
 View more versions of this product
Catalog No. 13-900-302
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
4 x 4.7 x 1.5 in. (10.16 x 11.9 x 3.8cm)
SBS
Aluminum Alloy
2 lb.
3.8cm
10.16cm
11.9cm
Ideal for all wet ice cooling, rapid snap-freezing in dry ice without alcohol
Twenty four 12 x 75mm FACS tubes
CoolRack™ XT CFT24
1kg
1.5 in.
4 in.
4.7 in.