Learn More
BioCision™ Automation-Friendly Sample Modules for CoolBox™ XT/2XT Systems
Thermo-conductive tube holders with SBS-footprint modules for easy transfer to robotic platforms
Manufacturer:  BioCision™ BCS537
 View more versions of this product
Catalog No. 13-900-306
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
5.2 x 3.5 x 0.7 in. (13.2 x 8.8 x 1.7cm)
SBS
Aluminum Alloy
1.7 lb.
1.7cm
13.2cm
8.8cm
Ideal for all wet ice cooling, rapid snap-freezing in dry ice without alcohol
One 96-well u-bottom plate
CoolRack™ XT 96U
0.8kg
0.7 in.
5.2 in.
3.5 in.