Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
BioCision™ Automation-Friendly Sample Modules for CoolBox™ XT/2XT Systems
Thermo-conductive tube holders with SBS-footprint modules for easy transfer to robotic platforms
Manufacturer:  BioCision™ BCS538
 View more versions of this product
Catalog No. 13-900-301
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
4 x 4.7 x 1.5 in. (10.16 x 11.9 x 3.8cm)
SBS
Aluminum Alloy
2 lb.
3.8cm
10.16cm
11.9cm
Ideal for all wet ice cooling, rapid snap-freezing in dry ice without alcohol
One 384-well PCR plate
CoolRack™ XT PCR384
1kg
1.5 in.
4 in.
4.7 in.