Learn More
BioCision™ CoolBox™ 2XT System
Benchtop ice-free cooling system requires no ice, electricity, or batteries
Manufacturer:  BioCision™ BCS503G
 View more versions of this product
Catalog No. 13-900-756
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Green
3, 4, or 5mL cryovials, 15mL or 50mL centrifuge tubes and blood collection tubes
13.4cm
15cm
10.5 in.
26.5cm
5 lb.
5.6 in.
20cm
10.5 x 7.8 x 6 in. (26.5 x 20 x 15cm)
Ideal for transporting samples from lab to lab, or field to clinic
5.27 in.
6 in.
Double capacity base, collar, lid and two XT cooling cores
8.26 in.
21cm
2.2kg
14.3cm
7.8 in.
2XT Cryogenic System