Learn More
BioCision™ CoolBox™ XT System
Benchtop ice-free cooling system requires no ice, electricity, or batteries
Manufacturer:  BioCision™ BCS502
 View more versions of this product
Catalog No. 13-900-758
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Purple
3, 4, or 5mL cryovials, 15mL or 50mL centrifuge tubes and blood collection tubes
13.4cm
15cm
7.87 in.
20cm
3 lb.
4.92 in.
16cm
7.87 x 6.2 x 6 in. (20 x 16 x 15cm)
Ideal for transporting samples from lab to lab, or field to clinic
5.27 in.
6 in.
Foam base, collar, lid and one XT cooling core
6.4 in.
16.5cm
1.36kg
12.5cm
6.2 in.
CoolBox™ XT System