BioCision™ CoolCell™ FTS30 Cell Freezing Container

High capacity, -1°C/minute cell freezing container with removable vial module

Manufacturer: BioCision™ BCS170G

 View more versions of this product

Catalog No. 13-900--680

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type Cell Freezing Container
    For Use With (Application) Provides standardized cryopreservation for all cells including cell lines, stem cells, blood and other primary cells and yeast
    Material HDPE, Foam
    Weight (Metric) 0.5kg
    Weight (English) 1 lb.
    Color Green
    Height (English) 4.5 in.
    Holds 30 1or 2mL cryogenic tubes
    Height (Metric) 11.43cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit