Learn More
BioCision™ CoolCell™ FTS30 Cell Freezing Container
High capacity, -1°C/minute cell freezing container with removable vial module
Manufacturer:  BioCision™ BCS170O
 View more versions of this product
Catalog No. 13-900--681
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Orange
4.5 in.
Provides standardized cryopreservation for all cells including cell lines, stem cells, blood and other primary cells and yeast
30 1or 2mL cryogenic tubes
1 lb.
11.43cm
Cell Freezing Container
HDPE, Foam
0.5kg