BioCision™ CoolCell™ LX 2mL Cryo Vial Freezers

Alcohol-free CoolCell LX delivers unparalleled cell freezing reproducibility

Manufacturer: BioCision™ BCS405G

 View more versions of this product

Catalog No. 13-900-857

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Material HDPE, Foam
    Holds 12 1 or 2mL cryogenic tubes
    Product Type Cell Freezing Container
    For Use With (Application) Ideal for all cells including cell lines, stem cells and primary cells
    Includes Beveled lid
    Design Solid core
    Color Green
    Height (Metric) 10.9cm
    Height (English) 4.2 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit