Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioCision™ CoolCell™ LX 2mL Cryo Vial Freezers

Alcohol-free CoolCell LX delivers unparalleled cell freezing reproducibility

Manufacturer:  BioCision™ BCS405PK

 View more versions of this product

Catalog No. 13-900-859

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications Description & Specifications

Specifications

Solid core
10.9cm
Cell Freezing Container
HDPE, Foam
12 1 or 2mL cryogenic tubes
Pink
4.2 in.
Ideal for all cells including cell lines, stem cells and primary cells
Beveled lid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit