Learn More
BioCision™ CoolProbe™ Traceable Temperature Monitor
Universal probe can be placed in tubes or plates
Manufacturer:  BioCision™ BCS200
 View more versions of this product
Catalog No. 13-900-686
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
±1°C
4.25 x 2.7 x 1.9 in. (108 x 70 x 48mm)
CoolRack™ and CoolSink™ modules
48mm
108mm
-58° to +158°F
1 lb.
2.7 in.
NIST
LCD
1.9 in.
4.25 in.
1°C
-50° to +70°C
0.45kg
70mm