Learn More
BioCision™ Hinged Cryoboxes
Standardizes cryogenic sample storage
Manufacturer:  BioCision™ BCS206P
 View more versions of this product
Catalog No. 13-901-104
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Purple
2 in.
5.25 in.
5.25 in.
Cryogenic Storage Box
Laminated Cardboard
81 1mL or 2mL cryogenic vial and microfuge tubes
5cm
13.3cm
13.3cm
5.25 x 5.25 x 2 in. (13.3 x 13.3 x 5cm)
Ideal for repeated freezing and thawing, ultra-low freezer storage or liquid nitrogen vapor
Hinged lid, 81-place adjustable plastic grid
5/Pk.