Learn More
BioCision™ TruCool™ Hinged Cryoboxes with 81-Place Grid
Standardizes cryogenic sample storage
Manufacturer:  BioCision™ BCS215G
 View more versions of this product
Catalog No. 13-901-158
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Green
3.5 in.
5.25 in.
5.25 in.
Cryogenic Storage Box
Plastic 81-place grid with adjustable slats for multiple vial types
8.9cm
13.3cm
13.3cm
5.25 x 5.25 x 3.5 in. (13.3 x 13.3 x 8.9cm)
Laminated Cardboard
5/Pk.