Learn More
BioCision™ Multipurpose Ice Buckets with Lids
Round buckets for ice, dry ice or other temperature media
Manufacturer:  BioCision™ BCS11525G
 View more versions of this product
Catalog No. 13-900-745
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Green
4.7 in.
9.5 in.
24.1cm
EVA Foam
Round
12.1cm
2.5L
Ice Bucket
Ideal for use with ice, dry ice, liquid nitrogen, saline solutions, or acetone
Lid