Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioCision™ TruCool™ Hinged Cryoboxes with 81-Place Grid

Standardizes cryogenic sample storage

Specifications

Type Cryo Box
Dimensions (L x W x H) 5.25 x 5.25 x 3.5 in. (13.3 x 13.3 x 8.9 cm)
Includes Plastic 81-Place Grid with Adjustable Slats
Material Fiberboard
View More Specs

Description

Description

  • Plastic 81-place grid for maximum storage capacity
  • Hinged lid ensures markings and samples remain in synch
  • Easy open lid minimizes contamination
  • Laminated waterproof fiberboard
  • Stores up to 81 cryogenic vials (5mL) or microtubes
  • Designed for repeated freezing and thawing
  • Bright colors are quickly visible and allow for sample segregation
  • L x W x H: 13.3 x 13.3 x 9.2cm (5.25 x 5.25 x 3.5 in.)
Specifications

Specifications

Cryo Box
5.25 x 5.25 x 3.5 in. (13.3 x 13.3 x 8.9 cm)
Plastic 81-Place Grid with Adjustable Slats
Fiberboard
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit