Learn More
Ansell™ BioClean-D™ Sterile Drop-Down Cleanroom Coveralls with Hood
BioClean-D™ Sterile Drop-Down Cleanroom Coveralls with Hood include internal colored tabs to indicate safe touch points, strategically placed removable tabs, and a removable zip tab.
Manufacturer:  Ansell™ SBDSH5XL
 View more versions of this product
Catalog No. 19-500-813
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • ISO Class 4 and EU GMP Grade A compatible
  • Antistatic, low linting durable CleanTough™ fabric
  • Quick release press stud tabs for touch-free donning
  • Elasticated back, hood, cuffs and ankles
  • Zip front
  • ASTM D6978 05 Tested
  • HELMKE DRUM Tested
  • Low-Particulate
  • Sterility Assurance Level 10-6
Specifications
Specifications
Elastic
Safety
ISO 9001:2015, (EU)2016/425, ASTM F739-12, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2, ISO 11137-1:2006
White
Quick release tabs, removable zip tab
Cleanroom, Pharma, Controlled Environments, PPE, 503B Facilities, Manufacturing, Industrial
Hooded
Bound, Stitched
Sterile
CleanTough™ fabric
Elastic
Zipper
Elastic
Yes
Elastic
Cleanroom coverall
5X-Large