Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask

Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology. Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask is ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.

$536.00 - $566.00

Specifications

Closure Material Phenolic
For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Neck Style Canted
Shape Rectangular
Surface Coating Collagen IV
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (Metric) Culture Area Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Culture Area Price Quantity & Availability  
08-774-35
View Documents
Corning™
354523
250 mL 75 cm2
Case of 10 for $536.00
Add to cart
 
08-774-36
View Documents
Corning™
354528
750 mL 175 cm2
Case of 5 for $566.00
Add to cart
 
08-774-37
Accessories View Documents
Corning™
354534
70 mL 25 cm2 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

 

  • BioCoat™ provides a superior solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures
  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

 

For research use only.

Specifications

Specifications

Phenolic
Canted
Collagen IV
Plug Seal
For adhesion, growth and differentiation of neuronal and transfected cell lines
Rectangular
Sterile
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit