Learn More
Aat BioquestSupplier Diversity Partner Biocytin C2 maleimide DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 3085
Biocytin maleimide readily reacts with thiol moieties of biopolymers to form thioether conjugate that is quite stable. This biocytin maleimide requires mild conjugation conditions.
Catalog No. 50-195-3217