Biogel™ Powder-Free Latex Diagnostic Gloves
Manufacturer: MOLNLYCKE 30385
 View more versions of this product
Catalog No. 18-999-4592G
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Coating Material Biogel™ Hydrogel Polymer
    Color Natural
    Glove Cuff Style Beaded
    Hand Hand-specific
    Length (English) 10.5 in.
    Length (Metric) 27cm
    Material Latex
    Powdered No
    Size 8.5
    Sterile No
    With patented Biogel coating. Low residual allergen levels specially processed to ensure <50µg of water extractable protein per gram of latex. Recommended for moderate- to high-risk exposures. Easy to don with damp or dry hands. 100% physically inspected. Provide outstanding comfort, fit, and feel. Beaded cuff and curved finger design help prevent rolldown and hand fatigue. Hand specific. Single use. Color: natural. Nonsterile.
  • Description & Specifications
Related Products
Product Content Feedback