Biogel™ Powder-Free Latex Diagnostic Gloves
Manufacturer: MOLNLYCKE 30390
 View more versions of this product
Catalog No. 18-999-4592H
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Powdered No
    Size 9
    Sterile No
    With patented Biogel coating. Low residual allergen levels specially processed to ensure <50µg of water extractable protein per gram of latex. Recommended for moderate- to high-risk exposures. Easy to don with damp or dry hands. 100% physically inspected. Provide outstanding comfort, fit, and feel. Beaded cuff and curved finger design help prevent rolldown and hand fatigue. Hand specific. Single use. Color: natural. Nonsterile.
  • Description & Specifications
Related Products
Product Content Feedback