Learn More
Thermo Scientific BioGold Microarray Slides, gold coated
Improve adhesion of biologically active cells with Thermo Scientific BioGold Microarray Slides' thin-film pure gold coating.
Manufacturer: Thermo Scientific C095076M20
Description
Improve adhesion of biologically active cells with Thermo Scientific BioGold Microarray Slides' thin-film pure gold coating. Manufactured from ultra flat, low-fluorescence glass; 800A (80 nm) of 99.999% pure gold over inert binding layer, these slides offer background reflective properties that provide significant optical advantages for detecting each cell's unique light-wave signature.
- 83% Reflectance at 543nm wavelength
- Printable area of 25 x 75mm
- Compatible with any microarray printer or scanner accommodating the industry standard
- Secure no-contact mBox packaging, vacuum sealed in a foil barrier bag
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Gold | |
Gold Coated Microarray Slides | |
25mm |
Fluorescence Glass | |
75mm | |
Plain |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.