Learn More
Sartorius™ Biohit™ PICUS™ 12-Channel Electronic Pipet - PROMO
Reduces risk of repetitive strain injury. Sartorius™ Biohit™ PICUS™ 12-Channel Electronic Pipet ensures the most accurate, precise and reliable results due to advanced DC-motor concept and built-in error control.
$1281.00
Specifications
Number of Channels 12
Finger Style Ergonomic
Warranty 2 Years
View More Specs
Products 4
Catalog Number Mfr. No. Color Volume (Metric) Increments Precision Price Quantity & Availability  
Catalog Number Mfr. No. Color Volume (Metric) Increments Precision Price Quantity & Availability  
14-559-360PR
View Documents
Sartorius™
735421PR
Gray 0.2 to 10 μL 0.02μL 3.00 to 0.50
Each for $1,281.00
Add to cart
14-559-361PR
View Documents
Sartorius™
735441PR
Yellow 5 to 120 μL 0.1μL 1.50 to 0.20
Each for $1,281.00
Add to cart
14-559-362PR
View Documents
Sartorius™
735461PR
Orange 10 to 300 μL 0.2μL 1.00 to 0.20
Each for $1,281.00
Add to cart
14-559-363PR
View Documents
Sartorius™
735491PR
Purple 50 to 1200 μL 1μL 1.00 to 0.20
Each for $1,281.00
Add to cart
Description
Description
Light and small electronic pipet ensures an optimal working posture and offers comfortable pipeting. New ergonomic design protects from RSI (repetitive strain injury) and eases workload in long pipetting series. Provides more reliable results with a new generation technology and error control system.
  • Helps prevent repetitive strain injury (RSI) due to lightweight, ergonomic design and electronic tip ejection
  • Ensures the most accurate, precise and reliable results due to advanced DC-motor concept and built-in error control
  • Saves time with extremely fast one-hand adjustment, a unique built-in microplate tracker and easy programmability
  • Adjustment wheel for scrolling menus, adjusting volume and controlling manual pipetting
  • Variety of pipetting modes in user friendly interface including memory places for favorite programs
  • Unique Tracker in conjunction with pipetting, reverse pipetting and multi dispensing modes to help microplate work
  • Adjustment possible for 1-point, 2-points and 3-points
  • Easy maintenance of lower parts with GLP information integrated in software for service and calibration
  • Safe-Cone filters in all models >10µL to prevent contamination
  • Autoclavable lower parts in all models (excluding 1200µL model)
  • Compatible with Biohit™ electronic pipet charging systems, charging stand for one pipet and charging carousel for four pipets
  • Direct charging option with micro USB cable
Specifications
Specifications
12
Ergonomic
2 Years