Learn More
Sartorius™ Biohit™ Picus™ NxT Electronic Pipettes, Single Channel PROMO
Designed to ease user's workload and to protect from repetitive strain injury (RSI)
Manufacturer:  Sartorius™ LH745111PR
 View more versions of this product
Catalog No. 14-557-826PR
Add to cart
Description
Description
Laboratory work requires repetition of similar pipetting steps that may cause strain on hand and arm, and thus may compromise results. To minimize this risk, a highly ergonomic electronic pipette should be used for minimum effort and strain. Sartorius Picus™ NxT are the most sophisticated and ergonomic electronic compact and light weight pipettes.
  • Password protection for stored programs
  • Very last liquid droplets dispensed by repeated blow-out for maximum recovery
  • Features one-touch operating-button and feather-light electronic tip ejection
  • Includes plate tracker for microwell plate dispensing
  • Electronic piston control and brake for precise pipetting
  • Safe-cone filters to prevent contamination
  • Adjustment wheel for volume setting and navigation
  • Range of pipetting modes for diluting and titrating to serial dispensing
  • Intuitive user interface in a choice of language options: English, French, German, Russian and Chinese
  • Certificate of accredited 3-point calibration
  • RFID-enabled to support asset management
Applications:
PCR and other DNA/RNA techniques, ELISA, Protein analysis and Cell culture
Specifications
Specifications
Single
Certificate of accredited 3-point calibration
Red
0.5 to 10 mL
Documents
Promotions
Promotions
  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021