Learn More
Sartorius™ Biohit™ SafetySpace™ Filter Pipet Tips PROMO
Sartorius™ Biohit™ SafeySpace™ Filter Pipet Tips allow pipetting any type of liquid of using any pipetting technique without the risk of the sample absorbing into the filter.
Manufacturer:  Sartorius™ 791001FPMO
 View more versions of this product
Catalog No. 14-559-511PMO
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Biohit™ SafetySpace™ Filter Tips leave more space between the sample and the filter than conventional filter tips. This allows pipetting any type of liquid or using any pipetting technique without the risk of the precious sample absorbing into the filter. In effect, this secures 100% sample recovery and thus greater accuracy.
  • Polypropylene
  • With regular air gap and filter barrier
  • Fully autoclavable
  • Single tray packaging
  • Purity level: RNase, DNase, Pre-endotoxin-free sterilized
The safety space feature is particularly useful in:
  • pipetting foaming liquids such as buffers and proteins
  • pipetting solvents
  • multiple dispensing functions of electronic pipettes
  • reverse pipetting
  • any other application requiring extra space in the tip
The tips are made of virgin polypropylene and the filter of polyethylene without self-sealing additives to avoid any interference with the sample and the results. The filter barrier helps protect against cross-contamination of the sample and/or the pipette and reduces the need for pipette maintenance. The filter tips are available in multiple volumes from 10 µl to 1200 µl and are packed in colour coded single tray racks. They are DNase, RNase and endotoxin free certified and pre-sterilized.
The filter barrier and the certified purity makes the filter tips ideal for:
  • molecular biology
  • microbiology
  • cell culture applications
  • radioactive work
Specifications
Specifications
Sartorius™ Biohit™, Universal Fit Pipettor
5 to 1000μL
Sterile
No
3.07 in.
CE, ISO, IVD
Sartorius™ Biohit™
Rack
Filtered
Blue
PP (Polypropylene)
7.8cm
DNase, RNase, and Endotoxin Free
Pre-sterilized
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021