Thermo Scientific BIOLiner™ Rotor Bucket Adapters
Combine Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters with the BIOLiner Rotor Bucket for a complete system.
Manufacturer: Thermo Scientific 75003692
 View more versions of this product
Catalog No. 75-003-692
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Description 750mL Flat-bottom Bottle w/Round Adapters for TX-750 Round Buckets
    Combine Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters with the BIOLiner Rotor Bucket for a complete system.
    Ordering Information:
    Individual adapters and bottle/adapter packages are offered.
    • Capacity range: 3L to 980mL
    • Maximum tube dimensions (O.D. x L): 98 x 124mm
    Includes:
    75-003-692 includes 750mL flat-bottom bottle with all round adapters for the TX-750 round bucket and four 500mL Corning™ conical bottle adapters.
  • Description & Specifications
Related Products
Product Content Feedback