Learn More
Thermo Scientific™ BIOLiner™ Swinging-Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Spin samples at 3,500rpm with the Thermo Scientific™ BIOLiner™ Swinging-Bucket Rotor.
Manufacturer:  Thermo Scientific™ 75003689
 View more versions of this product
Catalog No. 75-003-689
  Check Availability
Add to cart
Specifications
Description
Spin samples at 3,500rpm with the Thermo Scientific™ BIOLiner™ Swinging-Bucket Rotor.
Ordering Information:
Order BIOLiner Buckets (75-003-668, set of 4) and Thermo Scientific™ ClickSeal™ Biocontainment Lids (75-003-669, set of four) to complete assembly. Replacement O-rings for lids (75-003-688) and adapters are sold separately. Microplate carriers (50-036-70) and lids (75-003-689) are also available. Contact your sales representative for information.
Compatible with:
Thermo Scientific™ Sorvall™ ST40/40R, SL40/40R, Sorvall Legend™ XT/XTR, Heraeus™ Megafuge™ 40, Heraeus Multifuge™ X3/X3R, Heraeus Megafuge 40/40R Centrifuge Series
Description & Specifications
Specifications
90°
13307
3,500rpm
Thermo Scientific™ Sorvall™ Legend™ XT/XF, Sorvall ST 40, Heraeus™ Multifuge™ X3/X3F, Heraeus™ Megafuge™ 40 and SL 40/40F centrifuges
Sealing Lids for Microplate Carriers (Set of 4)
Sealing Lids for Microplate Carriers (set of 4)
Biocontainment certified by CAMR?, HPA, Porton Down, U.K.
4 x 750mL
2,739 x g
Swinging bucket rotor
Max./Min.: 20/10.5cm
98mm
124mm
Accessories