Learn More
Braintree ScientificSupplier Diversity Partner BIOLITE STAND DFS Item
Manufacturer:  Braintree ScientificSupplier Diversity Partner RIS200
Rodent Intubation Stand, this platform provides the perfect stage for intubating mice and rats up to 300 grams. The animal is suspended from the incisor wire on the angled side of the stand.
Catalog No. NC0065923
Due to product restrictions, please to purchase or view availability for this product.