Learn More
Carestream Health, Inc BioMax MR Film | 8 x 10in | 50/pack  DFS Item
Manufacturer:  Carestream Health, Inc 870 1302
BIOMAX MR is a single emulsion film that provides maximum resolution and sensitivity for detection of 35 S, 33 P, and 14 C labeled samples. BIOMAX MR enables improved scanning for clearer autoradiograms and offers improved detection using our “T-grain” emulsion technology. It can be processed manually or automatically, and is suitable for chemiluminescent detection.
Catalog No. NC1884694
Due to product restrictions, please to purchase or view availability for this product.