Learn More
Biomedical Polymers LeukoChek Leukocyte Test Kit
Complete test kit for the microscopic counting of leukocytes and platelets in whole blood
Manufacturer: Biomedical Polymers BMPLUKCHK50
Includes: Reservoir and capillary assembly
Description
Designed to replace the discontinued BD Unopette
- Leukocyte and platelet count procedures are performed exactly the same as with the BD Unopette (365854/365855)
- Independently tested and confirmed to produce equivalent results to the discontinued BD Unopette
- Tested to CLIA guidelines
- Full Validation Study available
Specifications
Microscopic Counting of Leukocytes and platelets in Whole Blood | |
Reservoir and capillary assembly | |
Leukocyte Test Kit | |
50 Pack |