Learn More
Aniara Diagnostica LLCSupplier Diversity Partner BIOPHEN UFH CTRL PLASMA 12/PK DFS Item
Manufacturer:  Aniara Diagnostica LLCSupplier Diversity Partner A223101
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. This Product is CE Marked. Health Canada Licensed. 510(k) Cleared for In Vitro Diagnostic Use. Quality control plasmas (2 different levels (about 0.20 and 0.50 IU/mL)) for the quality control of UFH testing (Unfractionated Heparin). The exact concentrations and the acceptance ranges are indicated on the flyer provided with each kit.
Catalog No. NC9782292