Learn More
Alere Triage™ Drugs of Abuse Panel
Immunoassay for the qualitative determination of the presence of parent compound and/or major metabolites of up to nine distinct drug classes in urine
Manufacturer:  Biosite, Inc. 90008
 View more versions of this product
Catalog No. 13-900-08
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Moderate
25 Tests
Room temperature
Triage™ drugs of abuse 8 panel
Amphetamines, Methamphetamines, Barbiturates, Benzodiazepines, Cocaine, Marijuana, Methadone, Opiates, PCP, Propoxyphene and Tricyclic Antidepressants
Urine
10 min.