Learn More
Biosynergy HemoCool™ Gel-Pak
For maintaining the temperature of blood bag during labeling and processing
Manufacturer:  Biosynergy 8100
Catalog No. 22-246-199
Add to cart
Description
Description
  • Reusable
  • Used for transport
Specifications
Specifications
Maintaining temperature of the blood bag/materials during labeling,testing
Gel Pack